missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ebola Virus Ebola Sudan Protein
A cDNA sequence encoding the Ebola Sudan Protein was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | Ebola Sudan Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Ebola Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938158
|
enQuireBio™
QP11726-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958158
|
enQuireBio™
QP11726-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948158
|
enQuireBio™
QP11726-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Ebola Sudan Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Ebola Virus | |
Untagged | |
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ebola Sudan protein solution is supplied in Phosphate buffer and 0.02% sodium azide. |