missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli UNG Protein

A cDNA sequence encoding the UNG was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13895-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965049

  • £5741.17 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

947067
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
ung
E. coli
His
UNG protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
UNG Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMANELTW HDVLAEEKQQ PYFLNTLQTV ASERQSGVTI YPPQKDVFNA FRFTELGDVK VVILGQDPYH GPGQAHGLAF SVRPGIAIPP SLLNMYKELE NTIPGFTRPN HGYLESWARQ GVLLLNTVLT VRAGQAHSHA SLGWETFTDK VISLINQHRE GVVFLLWGSH AQKKGAIIDK QRHHVLKAPH PSPLSAHRGF FGCNHFVLAN QWLEQRGETP IDWMPVLPAE SE
Videos
SDS
Documents
Certificates
Special Offers

Special Offers