missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli UDP-3-O-acyl-N-acetylglucosamine deacetylase Protein
A DNA sequence encoding the Escherichia coli (strain K12) UDP-3-O-acyl-N-acetylglucosamine deacetylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7317-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_414638.1 | |
UDP-3-O-acyl-N-acetylglucosamine deacetylase Protein | |
Research Use Only | |
lpxC | |
Recombinant Protein | |
E. coli | |
MIKQRTLKRI VQATGVGLHT GKKVTLTLRP APANTGVIYR RTDLNPPVDF PADAKSVRDT MLCTCLVNEH DVRISTVEHL NAALAGLGID NIVIEVNAPE IPIMDGSAAP FVYLLLDAGI DELNCAKKFV RIKETVRVED GDKWAEFKPY NGFSLDFTID FNHPAIDSSN QRYAMNFSAD AFMRQISRAR TFGFMRDIEY LQSRGLCLGG SFDCAIVVDD YRVLNEDGLR FEDEFVRHKM LDAIGDLFMC GHNIIGAFTA YKSGHALNNK LLQAVLAKQE AWEYVTFQDD AELPLAFKAP SAVLA | |
Greater than 80% as determined by SDS-PAGE. |
944816 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |