missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli tsaE Protein

A DNA sequence encoding the Escherichia coli (strain K12) tsaE, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7334-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048280

  • £628.00 / 100µg

Estimated Shipment: 29-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_418589.1
tsaE Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
948684
Greater than 90% as determined by SDS-PAGE.
Research Use Only
tsaE
Recombinant Protein
E. coli
MMNRVIPLPD EQATLDLGER VAKACDGATV IYLYGDLGAG KTTFSRGFLQ ALGHQGNVKS PTYTLVEPYT LDNLMVYHFD LYRLADPEEL EFMGIRDYFA NDAICLVEWP QQGTGVLPDP DVEIHIDYQA QGREARVSAV SSAGELLLAR LAG
Videos
SDS
Documents
Certificates