missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Thiosulfate sulfurtransferase GlpE Protein
A DNA sequence encoding the Escherichia coli (strain ATCC 8739 / DSM 1576 / Crooks) Thiosulfate sulfurtransferase GlpE, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7505-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000371929.1 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Thiosulfate sulfurtransferase GlpE Protein | |
Research Use Only | |
glpE | |
Recombinant Protein | |
Yeast | |
MDQFECINVA DAHQKLQEKE AVLVDIRDPQ SFAMGHAVQA FHLTNDTLGA FMRDNDFDTP VMVMCYHGNS SKGAAQYLLQ QGYDVVYSID GGFEVWQRQF PAEVAYGA | |
Greater than 90% as determined by SDS-PAGE. |