missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli S-ribosylhomocysteine lyase Protein

A DNA sequence encoding the Escherichia coli (strain K12) S-ribosylhomocysteine lyase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7508-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049080

  • £2480.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417172.1
S-ribosylhomocysteine lyase Protein
Research Use Only
luxS
Recombinant Protein
E. coli
PLLDSFTVDH TRMEAPAVRV AKTMNTPHGD AITVFDLRFC VPNKEVMPER GIHTLEHLFA GFMRNHLNGN GVEIIDISPM GCRTGFYMSL IGTPDEQRVA DAWKAAMEDV LKVQDQNQIP ELNVYQCGTY QMHSLQEAQD IARSILERDV RINSNEELAL PKEKLQELHI
Greater than 90% as determined by SDS-PAGE.
947168
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers