missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Penicillin-binding protein 1A Protein

A DNA sequence encoding the Escherichia coli (strain K12) Penicillin-binding protein 1A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7405-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048700

  • £628.00 / 100µg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417855.4
Penicillin-binding protein 1A Protein
100 μg
Not determined.
Partial (see sequence information for more details).
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
947907
Greater than 80% as determined by SDS-PAGE.
Research Use Only
mrcA
Recombinant Protein
E. coli
MLDEGYITQQ QFDQTRTEAI NANYHAPEIA FSAPYLSEMV RQEMYNRYGE SAYEDGYRIY TTITRKVQQA AQQAVRNNVL DYDMRHGYRG PANVLWKVGE SAWDNNKITD TLKALPTYGP LLPAAVTSAN PQQATAMLAD GSTVALSMEG VRWARPYRSD TQQGPTPRKV TDVLQTGQQI WVRQVGDAWW LAQVPEVNSA LVSINPQNGA VMALVGGFDF NQSKFNRATQ ALRQVGSNIK PFLYTAAMDK GLTLASMLND VPISRWDASA GSDWQPKNSP PQYAGPIRLR QGLGQSKNVV M
Videos
SDS
Documents
Certificates