missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli NTH Protein

A cDNA sequence encoding the NTH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12892-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961869

  • £99.15 / 2µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

947122
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MNKAKRLEIL TRLRENNPHP TTELNFSSPF ELLIAVLLSA QATDVSVNKA TAKLYPVANT PAAMLELGVE GVKTYIKTIG LYNSKAENII KTCRILLEQH NGEVPEDRAA LEALPGVGRK TANVVLNTAF GWPTIAVDTH IFRVCNRTQF APGKNVEQVE EKLLKVVPAE FKVDCHHWLI LHGRYTCIAR KPRCGSCIIE DLCEYKEKVD I
Greater than 85.0% as determined by SDS-PAGE.
NTH Protein
Research Use Only
nth
E. coli
His
The NTH solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT, 0.1mM PMSF and 40% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers