missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli MSRB Protein

A cDNA sequence encoding the MSRB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12743-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951399

  • £151.37 / 20µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MSRB Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
MSRB protein solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MANKPSAEEL KKNLSEMQFY VTQNHGTEPP FTGRLLHNKR DGVYHCLICD APLFHSQTKY DSGCGWPSFY EPVSEESIRY IKDLSHGMQR IEIRCGNCDA HLGHVFPDGP QPTGERYCVN SASLRFTDGE NGEEING
Videos
SDS
Documents
Certificates
Special Offers

Special Offers