missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli LptA Protein

A DNA sequence encoding the Escherichia coli (strain K12) LptA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7331-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048250

  • £715.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417667.1
LptA Protein
Research Use Only
lptA
Recombinant Protein
E. coli
VTGDTDQPIH IESDQQSLDM QGNVVTFTGN VIVTQGTIKI NADKVVVTRP GGEQGKEVID GYGKPATFYQ MQDNGKPVEG HASQMHYELA KDFVVLTGNA YLQQVDSNIK GDKITYLVKE QKMQAFSDKG KRVTTVLVPS QLQDKNNKGQ TPAQKKGN
Greater than 90% as determined by SDS-PAGE.
947920
100 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers