missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli glpE Protein
A cDNA sequence encoding the glpE was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11989-20ug
Additional Details : Weight : 0.01000kg
Specifications
glpE Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDQFECI NVADAHQKLQ EKEAVLVDIR DPQSFAMGHA VQAFHLTNDT LGAFMRDNDF DTPVMVMCYH GNSSKGAAQY LLQQGYDVVY SIDGGFEAWQ RQFPAEVAYG A | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
The glpE solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |