missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Ferric uptake regulation Protein

A DNA sequence encoding the Escherichia coli (strain K12) Ferric uptake regulation protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7394-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048640

  • £715.00 / 100µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_415209.1
Ferric uptake regulation Protein
Research Use Only
fur
Recombinant Protein
E. coli
TDNNTALKKA GLKVTLPRLK ILEVLQEPDN HHVSAEDLYK RLIDMGEEIG LATVYRVLNQ FDDAGIVTRH NFEGGKSVFE LTQQHHHDHL ICLDCGKVIE FSDDSIEARQ REIAAKHGIR LTNHSLYLYG HCAEGDCRED EHAHEGK
Greater than 90% as determined by SDS-PAGE.
945295
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers