Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 1 NS1 Protein
Recombinant Protein
Brand: enQuireBio™ QP10475-500ug
Additional Details : Weight : 0.01000kg
Description
The NS1, non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.Specifications
Serotype 1 NS1 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |
500 μg | |
Untested | |
Dengue virus (Strain: Nauru/Western Pacific/1974) | |
Native | |
>95% as determined by SDS-PAGE. |