missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue NS1 ST1 Protein
A cDNA sequence encoding the Dengue NS1 ST1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11644-10ug
Dettagli aggiuntivi : Weight : 0.01000kg
Specifica
Dengue NS1 ST1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue NS1 ST1 protein solution (1 mg/ml) in 1xPBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |