missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Epitope 13 Protein

A cDNA sequence encoding the Dengue Epitope 13 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11640-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937888

  • £583.00 / 500µg

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dengue Epitope 13 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
Untagged
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline, pH 7.4 and 0.02% sodium nitrate.
Videos
SDS
Documents