missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-3 Protein
A cDNA sequence encoding the Dengue Envelope-3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11631-2ug
Additional Details : Weight : 0.01000kg
Specifications
Dengue Envelope-3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue Envelope-3 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >90% pure as determined by 12.5% SDS-PAGE. |