missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant CMV CMV Pp65 Protein

A cDNA sequence encoding the CMV Pp65 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11465-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957348

  • £173.00 / 100µg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Pp65 Protein
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, 10mM glutathione, 60mM NaCl, 0.125% sarcosil and 50% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
CMV
GST
CMV Pp65 protein is >95% pure as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers