missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant CMV CMV Pp150 Protein
A cDNA sequence encoding the CMV Pp150 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1100.00
Specifications
Name | Pp150 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | CMV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967338
|
enQuireBio™
QP11462-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987338
|
enQuireBio™
QP11462-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977338
|
enQuireBio™
QP11462-1MG |
1 mg |
£1100.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Pp150 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
CMV | |
Untagged | |
CMV Pp150 protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 25mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol. |