missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chikungunya Virus CHIKV E1 Protein
A cDNA sequence encoding the CHIKV E1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | CHIKV E1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chikungunya Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987158
|
enQuireBio™
QP11405-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977158
|
enQuireBio™
QP11405-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967158
|
enQuireBio™
QP11405-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997158
|
enQuireBio™
QP11405-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907168
|
enQuireBio™
QP11405-EC-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CHIKV E1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chikungunya Virus | |
Untagged | |
Protein is 95% pure as determined by 12.5% SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
CHIKV E1 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, and Pepstatin A. |