missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Syntaxin-6 Protein

A DNA sequence encoding the Gallus gallus (Chicken) Syntaxin-6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6743-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095510

  • £715.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001006531.1
Syntaxin-6 Protein
Research Use Only
STX6
Recombinant Protein
E. coli
EDPFFVVKGE VQKAVNTAQG LFQRWTELLQ DPSIATREEI DWTTNELRNN LRSIEWDLED LDETISIVEA NPRKFNLDAT ELGIRKSFIT STRQVVREMK DQMSNSSMQA LAERKNRQAL LGESSSQSWS SGPDKYSRLD RELQLANSHF IEEQQAQQQL IVEQQDEQLE LVSGSIGVLK NMSQRIGGEL EEQAVMLDDF SHELDSTHSR LDNVMKKLAK VSHMTSDR
Greater than 90% as determined by SDS-PAGE.
424413
100 μg
Not determined.
Partial (see sequence information for more details).
Chicken
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers