missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Homeobox protein engrailed-1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Homeobox protein engrailed-1, was expressed in the hosts and tags indicated.
£292.00 - £2410.48
Specifications
Name | Homeobox protein engrailed-1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | EN1 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959129
|
enQuireBio™
QP5983-EC-10UG |
10 μg |
£292.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949129
|
enQuireBio™
QP5983-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999129
|
enQuireBio™
QP5983-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15979129
|
enQuireBio™
QP5983-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15989129
|
enQuireBio™
QP5983-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969129
|
enQuireBio™
QP5983-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Homeobox protein engrailed-1 Protein | |
Not determined. | |
Full Length | |
Chicken | |
His-B2M | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
EN1 | |
Recombinant Protein | |
E. coli | |
MEEPPEGHGH HRDAAPPGPA NGGGGGGGGS DGDSAPVSPS PAPASPAAPC PLPLPRRRPP PPPPPRTTNF FIDNILRPDF GCKKEPPAAT GAATGAGGGG GGGGREQRDG AQSAGRENVN PLLARPPHAP SSALLCPDSN CAPDGSAPAG TAAKANPGTA AGAAGAAGAA KAQGDGGETP AAKYGEHGSP AILLMGSNNG GAVLKPDSQQ PLVWPAWVYC TRYSDRPSSP RTRKLKKKKT EKEDKRPRTA FTAEQLQRLK AEFQANRYIT EQRRQSLAQE LSLNESRVKI WFQNKRAKIK KATGIKNGLA LHLMAQGLYN HSTTTVQDKE ESE | |
Greater than 90% as determined by SDS-PAGE. |