missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Homeobox protein engrailed-1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Homeobox protein engrailed-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5983-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Homeobox protein engrailed-1 Protein | |
Research Use Only | |
EN1 | |
Recombinant Protein | |
E. coli | |
MEEPPEGHGH HRDAAPPGPA NGGGGGGGGS DGDSAPVSPS PAPASPAAPC PLPLPRRRPP PPPPPRTTNF FIDNILRPDF GCKKEPPAAT GAATGAGGGG GGGGREQRDG AQSAGRENVN PLLARPPHAP SSALLCPDSN CAPDGSAPAG TAAKANPGTA AGAAGAAGAA KAQGDGGETP AAKYGEHGSP AILLMGSNNG GAVLKPDSQQ PLVWPAWVYC TRYSDRPSSP RTRKLKKKKT EKEDKRPRTA FTAEQLQRLK AEFQANRYIT EQRRQSLAQE LSLNESRVKI WFQNKRAKIK KATGIKNGLA LHLMAQGLYN HSTTTVQDKE ESE | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Chicken | |
His-B2M | |
Tris-based buffer, 50% glycerol |