missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Homeobox protein engrailed-1 Protein

A DNA sequence encoding the Gallus gallus (Chicken) Homeobox protein engrailed-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5983-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15949129

  • £715.00 / 100µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Homeobox protein engrailed-1 Protein
Research Use Only
EN1
Recombinant Protein
E. coli
MEEPPEGHGH HRDAAPPGPA NGGGGGGGGS DGDSAPVSPS PAPASPAAPC PLPLPRRRPP PPPPPRTTNF FIDNILRPDF GCKKEPPAAT GAATGAGGGG GGGGREQRDG AQSAGRENVN PLLARPPHAP SSALLCPDSN CAPDGSAPAG TAAKANPGTA AGAAGAAGAA KAQGDGGETP AAKYGEHGSP AILLMGSNNG GAVLKPDSQQ PLVWPAWVYC TRYSDRPSSP RTRKLKKKKT EKEDKRPRTA FTAEQLQRLK AEFQANRYIT EQRRQSLAQE LSLNESRVKI WFQNKRAKIK KATGIKNGLA LHLMAQGLYN HSTTTVQDKE ESE
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Chicken
His-B2M
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers