missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine LRG1 Protein
A cDNA sequence encoding the LRG1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12582-10ug
Additional Details : Weight : 0.01000kg
Specifications
611876 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Canine | |
MSPWSPERSQ SPGGLSSHLS KTLLLLLFSV ASAQGVTPDR DTCLVFPSSN GSSVSCHPPA KLPHHFPSDT VHLVVEFFNL TQLRPDTLQG VSNLQELHLS TNQLESLSPK LLLPVPLLKV LDLTRNALTQ LPPGLFRVSA ALHTLVLKEN RLEVLEPSWL CGLKALGHLD LSGNHLQTLP PGLLANVTAL RILDLSNNQL KTLPLDLLRG PLQLERLHLE GNRLQVLEEG LLAPQPGLRY LFLSDNKLAA VAAGAFRGLQ QLDMLDLSNN SLTSVPKGLW TSLGQPTRDM EDGFDISGNP WICDHNLDDL YRWLVANKDK MFSRNDTRCA RPEALKGQTL LEAAESH | |
Purity greater than 90% as determined by SDS-PAGE. |
LRG1 Protein | |
Research Use Only | |
LRG1 | |
Canine | |
Untagged | |
LRG1 protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 8.0. |