missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine IL6 Protein
A cDNA sequence encoding the IL6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12427-2ug
Additional Details : Weight : 0.01000kg
Specifications
403985 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
FPTPGPLAGD SKDDATSNSL PLTSANKVEE LIKYILGKIS ALRKEMCDKF NKCEDSKEAL AENNLHLPKL EGKDGCFQSG FNQETCLTRI TTGLVEFQLH LNILQNNYEG DKENVKSVHM STKILVQMLK SKVKNQDEVT TPDPTTDASL QAILQSQDEC VKHTTIHLIL RSLEDFLQFS LRAVRIMLEH HHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
IL6 Protein | |
Research Use Only | |
IL6 | |
Canine | |
His | |
IL6 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |