missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine GM CSF Protein
A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.
£99.15 - £3220.00
Specifications
Name | GM CSF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 as calculated by the dose-dependent stimulation of the proliferation of human TF1 erythroleukemic cells is typically 1-4 ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944518
|
enQuireBio™
QP10648-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924518
|
enQuireBio™
QP10648-10UG |
10 μg |
£160.00
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934518
|
enQuireBio™
QP10648-1MG |
1 mg |
£3220.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
GM CSF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APTRSPTLVT RPSQHVDAIQ EALSLLNNSN DVTAVMNKAV KVVSEVFDPEGPTCLETRLQ LYKEGLQGSL TSLKNPLTMM ANHYKQHCPP TPESPCATQNINFKSFKENL KDFLFNIPFD CWKPVKK | |
Greater than 96.0% as determined by1. Analysis by RP-HPLC. 2. Analysis by SDS-PAGE. |
Research Use Only | |
The ED50 as calculated by the dose-dependent stimulation of the proliferation of human TF1 erythroleukemic cells is typically 1-4 ng/ml. | |
Canine | |
Untagged | |
GMCSF was lyophilized after extensive dialysis against 1xPBS pH 7.4. |