missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type C1 Protein

A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type C1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7104-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077390

  • £780.00 / 100µg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Botulinum neurotoxin type C1 Protein
Research Use Only
Partial (see sequence information for more details).
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
100 μg
Not determined.
Recombinant Protein
E. coli
PITINNFNYS DPVDNKNILY LDTHLNTLAN EPEKAFRITG NIWVIPDRFS RNSNPNLNKP PRVTSPKSGY YDPNYLSTDS DKDPFLKEII KLFKRINSRE IGEELIYRLS TDIPFPGNNN TPINTFDFDV DFNSVDVKTR QGNNWVKTGS INPSVIITGP RENIIDPETS TFKLTNNTFA AQEGFGALSI ISISPRFMLT YSNATNDVGE GRFSKSEFCM DPILILMHEL NHAMHNLYGI AIPNDQTISS VTSNIFYSQY NVKLEYAEIY AFGGPTIDLI PKSARKYFEE KALDYYRSIA KRLNSITTAN PSSFNKYIGE YKQKLIRKYR FVVESSGEVT VNRNKFVELY NELTQIFTEF NYAKIYNVQN RKIYLSNVYT PVTANILDDN VYDIQNGFNI PKSNLNVLFM GQNLSRNPAL RKVNPEN
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers