missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine TROVE2 Protein
A cDNA sequence encoding the TROVE2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £4625.00
Specifications
Name | TROVE2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954809
|
enQuireBio™
QP13820-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934809
|
enQuireBio™
QP13820-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944809
|
enQuireBio™
QP13820-1MG |
1 mg |
£4625.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TROVE2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Greater than 60% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Bovine | |
TROVE2 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol. |