missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Thrombin Protein
A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.
£99.15 - £219.00
Specifications
Name | Thrombin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | 2000 NIH U/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945448
|
enQuireBio™
QP10879-100U |
100 IU |
£109.00
100 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955448
|
enQuireBio™
QP10879-250U |
250 U |
£219.00
250 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965448
|
enQuireBio™
QP10879-50U |
50 IU |
£99.15
50 units |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Thrombin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Bovine | |
Lyophilized freeze dry powder formulated with Tris buffer and sodium chloride. |
Research Use Only | |
2000 NIH U/mg. | |
Bovine | |
Untagged |