missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Resistin / ADSF / RETN Protein
A DNA sequence encoding the Bos taurus (Bovine) Resistin / ADSF / RETN, was expressed in the hosts and tags indicated.
£292.00 - £2410.48
Specifications
Accession Number | NP_899206.1 |
---|---|
Gene ID (Entrez) | 369020 |
Name | Resistin / ADSF / RETN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094440
|
enQuireBio™
QP6598-EC-10UG |
10 μg |
£292.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014450
|
enQuireBio™
QP6598-EC-200UG |
200 μg |
£1165.00
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024450
|
enQuireBio™
QP6598-EC-500UG |
500 μg |
£1605.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034450
|
enQuireBio™
QP6598-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084440
|
enQuireBio™
QP6598-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004450
|
enQuireBio™
QP6598-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_899206.1 | |
Resistin / ADSF / RETN Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
369020 | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
QSLCPIDKAI SEKIQEVTTS LVPGAVRIIG LDCRSVTSRG SLVTCPSGFA VTGCTCGSAC GSWDVRAETT CHCQCAGMDW TGARCCRLHI Q | |
Greater than 90% as determined by SDS-PAGE. |