missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine PI3Ka Protein

A cDNA sequence encoding the PI3Ka was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13049-2.5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962349

  • £280.03 / 2.50µg
Estimated Shipment: 14-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PI3Ka Protein
2.5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
PI3Ka solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers