missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine HTF Protein
Recombinant Protein
Brand: enQuireBio™ QP12351-150mg
Additional Details : Weight : 0.01000kg
Specifications
280705 | |
150 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRPAVRALLA CAVLGLCLAD PERTVRWCTI STHEANKCAS FRENVLRILE SGPFVSCVKK TSHMDCIKAI SNNEADAVTL DGGLVYEAGL KPNNLKPVVA EFHGTKDNPQ THYYAVAVVK KDTDFKLNEL RGKKSCHTGL GRSAGWNIPM AKLYKELPDP QESIQRAAAN FFSASCVPCA DQSSFPKLCQ LCAGKGTDKC ACSNHEPYFG YSGAFKCLME GAGDVAFVKH STVFDNLPNP EDRKNYELLC GDNTRKSVDD YQECYLAMVP SHAVVARTVG GKEDVIWELL NHAQEHFGKD KPDNFQLFQS PHGKDLLFKD SADGFLKIPS KMDFELYLGY EYVTALQNLR ESKPPDSSKD ECMVKWCAIG HQERTKCDRW SGFSGGAIEC ETAENTEECI AKIMKGEADA MSLDGGYLYI AGKCGLVPVL AENYKTEGES CKNTPEKGYL AVAVVKTSDA NINWNNLKDK KSCHTAVDRT AGWNIPMGLL YSKINNCKFD EFFSAGCAPG SPRNSSLCAL CIGSEKGTGK ECVPNSNERY YGYTGAFRCL VEKGDVAFVK DQTVIQNTDG NNNEAWAKNL KKENFEVLCK DGTRKPVTDA ENCHLARGPN HAVVSRKDKA TCVEKILNKQ QDDFGKSVTD CTSNFCLFQS NSKDLLFRDD TKCLASIAKK TYDSYLGDDY VRAMTNLRQC STSKLLEACT FHKP | |
Greater than 97.0% as determined by Cellulose Acetate. |
HTF Protein | |
Research Use Only | |
TF | |
Bovine | |
Untagged | |
The protein (1 mg/ml) was lyophilized with no additives. |