missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Complement C3 Protein
A DNA sequence encoding the Bos taurus (Bovine) Complement C3, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_001035559.2 |
---|---|
Gene ID (Entrez) | 280677 |
Name | Complement C3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049960
|
enQuireBio™
QP7640-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069960
|
enQuireBio™
QP7640-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059960
|
enQuireBio™
QP7640-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079960
|
enQuireBio™
QP7640-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089960
|
enQuireBio™
QP7640-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099960
|
enQuireBio™
QP7640-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035559.2 | |
Complement C3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
280677 | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
SDLDDDIIPE EDIISRSQFP ESWLWTVIED LKQADKNGIS TKLMNVFLKD SITTWEILAV SLSDKKGICV ADPYEVTVMQ DFFIDLRLPY SVVRNEQVEI RAILYNYREA ENLKVRVELL YNPAFCSLAT AKKRHQQTIT IPARSSVAVP YVIVPLKIGL HEVEVKAAVY NHFISDGVKK TLKVVPEGVR VNKTVAVRTL NPEHLGQ | |
Greater than 90% as determined by SDS-PAGE. |