missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Alkaline Phosphatase Protein
A cDNA sequence encoding the Alkaline Phosphatase was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10518-5mg
Additional Details : Weight : 0.01000kg
Specifications
Alkaline Phosphatase Protein | |
Research Use Only | |
>1000 U/mg (pH 9.6, 25C and 0.025M glycin, 10% glycerol as buffer. | |
Bovine | |
Untagged | |
50% glycerol, 5mM MgCl2, 0.1mM ZnCl2 and 5mM TRIS, pH 7.0. |
5 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Bovine | |
MQGACVLLLL GLHLQLSLGL VPVEEEDPAF WNRQAAQALD VAKKLQPIQT AAKNVILFLG DGMGVPTVTA TRILKGQMNG KLGPETPLAM DQFPYVALSK TYNVDRQVPD SAGTATAYLC GVKGNYRTIG VSAAARYNQC KTTRGNEVTS VMNRAKKAGK SVGVVTTTRV QHASPAGAYA HTVNRNWYSD ADLPADAQMN GCQDIAAQLV NNMDIDVILG GGRKYMFPVG TPDPEYPDDA SVNGVRKRKQ NLVQAWQAKH QGAQYVWNRT ALLQAADDSS VTHLMGLFEP ADMKYNVQQD HTKDPTLQEM TEVALRVVSR NPRGFYLFVE GGRIDHGHHD DKAYMALTEA GMFDNAIAKA NELTSELDTL ILVTADHSHV FSFGGYTLRG TSIFGLAPSK ALDSKSYTSI LYGNGPGYAL GGGSRPDVND STSEDPSYQQ QAAVPQASET HGGEDVAVFA RGPQAHLVHG VEEETFVAHI MAFAGCVEPY TDCNLPAPTT ATSIPDAAHL AASPPPLALL AGAMLLLLAP TLY | |
95% pure by Gel Filtration. |