missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine aFGF Protein
A cDNA sequence encoding the aFGF was constructed and used to recombinantly synthesize the protein.
£99.15 - £2075.00
Specifications
Gene ID (Entrez) | 281160 |
---|---|
Name | aFGF Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | FGF1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934088
|
enQuireBio™
QP10508-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944088
|
enQuireBio™
QP10508-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924088
|
enQuireBio™
QP10508-100UG |
100 μg |
£2075.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
281160 | |
Research Use Only | |
FGF1 | |
Recombinant Protein | |
Bovine | |
MAEGETTTFT ALTEKFNLPL GNYKKPKLLY CSNGGYFLRI LPDGTVDGTK DRSDQHIQLQ LCAESIGEVY IKSTETGQFL AMDTDGLLYG SQTPNEECLF LERLEENHYN TYISKKHAEK HWFVGLKKNG RSKLGPRTHF GQKAILFLPL PVSSD | |
Greater than 90%. |
aFGF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Stimulates growth of bovine capillary endothelial cells by 3-5 fold over 5% calf serum at 10-25ng/ml FGF and 0ug/ml heparin. | |
Bovine | |
Untagged | |
Each 5microg aFGF were lyophilized from 0.5ml solution containing 1mM sodium phosphate, pH 7 after filtration over a low binding membrane. |