missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacillus Levanase Protein

A DNA sequence encoding the Bacillus sp. (strain L7) Levanase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7478-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048940

  • £715.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Levanase Protein
Research Use Only
Partial (see sequence information for more details).
Bacillus
His-SUMO
Tris-based buffer, 50% glycerol
100 μg
Not determined.
Recombinant Protein
E. coli
LPWNDLGHVW SGSAVADTTN ASGLFGSSGG KGLIAYYTSY NPDRHNGNQK IGLAYSTDRG RTWKYSEEHP VVIENPGKTG EDPGGWDFRD PKVVRDEANN RWVMVVSGGD HIRLFTSTNL LNWTLTDQF
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers