Learn More
Hellma Recertification of Glass Filter Set
For checking photometric accuracy and wavelength accuracy.
Brand:  Hellma 666S006
Additional Details : Weight : 0.35000kg
 View more versions of this product
Product Code. 15960421
  • £1091.40 / 1 set
Estimated Shipment: 30-12-2021
Description
Description
  • Spectrophotometers must be checked and recertified at regular intervals.
  • In general, a recertification interval of 12 months is recommended for checking and recertifying glass filters during the first two years of use, with an interval of 24 months thereafter.
Specifications
Specifications
For checking photometric accuracy and wavelength accuracy
Set of glass filters: F0, F2, F3, F4
666-S006