missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rec 15/2615 dihydrochloride, Tocris Bioscience™
Selective α1B antagonist
Brand: Tocris (Bio-Techne) 328410
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 10mg |
---|
Description
Selective α1B-adrenoceptor antagonist (Ki values are 0.3, 1.9 and 2.6 nM at human α1B, α1A and α1D receptors respectively).Chemical Identifiers
173059-17-1 | |
568.496 | |
rec 15/2615 dihydrochloride, 1-4-amino-6,7-dimethoxy-2-quinazolinyl-4-2-methoxy-6-1-methylethyl phenoxy acetyl piperazine dihydrochloride | |
1-[4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-2-(2-methoxy-6-propan-2-ylphenoxy)ethanone;dihydrochloride |
C26H35Cl2N5O5 | |
XZGSTPYGKYGQLD-UHFFFAOYSA-N | |
56972225 | |
CC(C)C1=C(C(=CC=C1)OC)OCC(=O)N2CCN(CC2)C3=NC4=CC(=C(C=C4C(=N3)N)OC)OC.Cl.Cl |
Specifications
Rec 15/2615 dihydrochloride | |
173059-17-1 | |
rec 15/2615 dihydrochloride, 1-4-amino-6,7-dimethoxy-2-quinazolinyl-4-2-methoxy-6-1-methylethyl phenoxy acetyl piperazine dihydrochloride | |
CC(C)C1=C(C(=CC=C1)OC)OCC(=O)N2CCN(CC2)C3=NC4=CC(=C(C=C4C(=N3)N)OC)OC.Cl.Cl | |
568.496 | |
568.49 |
10mg | |
C26H35Cl2N5O5 | |
XZGSTPYGKYGQLD-UHFFFAOYSA-N | |
1-[4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl]-2-(2-methoxy-6-propan-2-ylphenoxy)ethanone;dihydrochloride | |
56972225 | |
>98% |