missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ RBP4 Recombinant Human Protein

Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10354H08H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13475826

  • £855.71 / 250µg


Estimated Shipment
08-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Measured by its ability to bind all-trans retinoic acid
  • Binding of retinoic acid results in quenching of tryptophan fluorescence in RBP4
  • Recombinant human RBP4 can bind >1.0μM all-trans retinoic acid
  • Lyophilized in PB
Specifications

Specifications

Analysis of protein structure and performing functional assays
5950
≥85% by SDS-PAGE
Human
<1 EU/μg
RBP7
Other proteins
RBP4 Recombinant Human Protein
Lyophilized
23kd
5 x 50μg
Recombinant
Retinol binding protein 4, plasma
Assay Standard
Mammalian
His Tag
Videos