missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ RBP4 Recombinant Human Protein

Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10354H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13485826

  • £57.00 / 5µg


Estimated Shipment
15-03-2018
Add to basket

Description

Description

  • Measured by its ability to bind all-trans retinoic acid
  • Binding of retinoic acid results in quenching of tryptophan fluorescence in RBP4
  • Recombinant human RBP4 can bind >1.0μM all-trans retinoic acid
  • Lyophilized in PB
Specifications

Specifications

Analysis of protein structure and performing functional assays
5950
≥85% by SDS-PAGE
Human
<1 EU/μg
RBP7
Other proteins
RBP4 Recombinant Human Protein
Lyophilized
23kd
5μg
Recombinant
Retinol binding protein 4, plasma
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.