Learn More
Invitrogen™ TNF alpha Rat ELISA Kit
For the quantitative detection of rat TNF-alpha.
Brand: Invitrogen™ BMS622TEN
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with polyclonal antibody to rat TNF-alpha
Biotin-Conjugate anti-rat TNF-alpha polyclonal antibody
Streptavidin-HRP
Rat TNF-alpha Standard lyophilized, 5ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
The Rat Tumor Necrosis Factor alpha(Rt TNFα) ELISA quantitates Rt TNFα in rat serum or cell culture medium. The assay will exclusively recognize both natural and recombinant rat TNFα. Principle of the method The Rat TNF&alph
Tumor Necrosis Factor is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF-alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF-alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF-alpha. Due to its varied actions throughout the immune system, TNF-alpha may play a role in the pathogenesis of many disease states. TNF-alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF-alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF-alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF-alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1.Specifications
P16599 | |
11.0 pg/mL | |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
RATTNF,TNF-alphaa,Tnf | |
5% | |
HRP | |
RUO | |
2°C to 8°C | |
Rat | |
3 hr. 10 min. |
39.1-2,500 pg/mL | |
Biotin | |
Serum, Supernatant | |
ELISA | |
24835 | |
10% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Serum, 50 μL; Supernatant, 50 μL | |
Cachectin, Tumor necrosis factor alpha, DIF | |
1 hr. 20 min. |
For Research Use Only