missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TGF beta-1 Rat ELISA Kit
For the quantitative detection of rat TGF-beta1.
Brand: Invitrogen™ BMS623-3
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to rat TGF-beta1
Biotin-Conjugate anti-rat TGF-beta1 polyclonal antibody
Streptavidin-HRP
Rat TGF-beta1 Standard lyophilized, 4000pg/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrat Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
TGF beta-1 (TGFB1, Transforming Growth Factor Beta 1) is a polypeptide member of the transforming growth factor beta superfamily of cytokines, found almost ubiquitously in tissues. Transforming growth factor (TGF)-b is stored in the extracellular matrix as a latent complex with its pro-domain. Activation of TGF beta-1 requires the binding of aV integrin to an RGD sequence in the prodomain and exertion of force on this domain, which is held in the extracellular matrix by latent TGF-b binding proteins. Latent forms are complexes of TGF beta-1, an amino-terminal portion of the TGF-beta precursor, designated TGF-LAP (TGF-latency associated peptide), and a specific binding protein, known as LTBP. TGF beta-1 helps regulates proliferation, differentiation, adhesion, migration in many cell types. Many cells have TGF beta receptors, and the protein positively and negatively regulates many other growth factors. TGF beta-1 is cleaved into a latency-associated peptide and a mature TGF beta-1 peptide, and is found in either a latent form composed of a TGFB1 homodimer, a LAP homodimer, and a latent TGFB1-binding protein, or in an active form composed of a TGF beta-1 homodimer. The mature peptide may also form heterodimers with other TGF beta family members. The gene for TGF beta-1 is frequently upregulated in tumor cells, and mutations in this gene result in Camurati-Engelmann disease and cystic fibrosis.Specifications
P17246 | |
7.8 pg/mL | |
ELISA Kit | |
Rat | |
Colorimetric Microplate Reader | |
Tgfb | |
6.8% | |
HRP | |
RUO | |
2°C to 8°C | |
Rat | |
4 hr. |
31.25-2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
59086 | |
6.8% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
Camurati-Engelmann disease, DPD1 | |
1 hr. 20 min. |