missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IL-6 ELISA Kits

Measure levels of rat interleukin-6 in rat serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ ER3IL6

Code : PN

Additional Details : Weight : 0.20000kg Transport : UN number : 2796 Chem class : 2a Pack group : II

 View more versions of this product

Product Code. 11582631

  • £427.32 / 1 set


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

These rat IL-6 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interleukin-6 (IL-6)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 62.5 to 4000pg/mL

  • Sensitivity: < 16pg/mL

  • Precision: Dependent upon sample type

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IL-10 and TNFα; mouse IL-6, IL-1β, TNFα, IL-4, IL-2 and IL-1α; or human IL-8 and GM-CSF

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

62.5 to 4000pg/mL
Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
20X Wash Buffer, 55mL
Absorbance at 450nm minus absorbance at 550nm
<16pg/mL
50µL
96 microplate wells
Natural and recombinant rat interleukin-6 (IL-6)
Rat IL-6 ELISA Kit
Complete ELISA kit with coated 96-well plate
Dependent upon sample type
1 plate kit
Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with rat IL-10 and TNFa; mouse IL-6, IL-1ß, TNFa, IL-4, IL-2 and IL-1a; or human IL-8 and GM-CSF
4 hr.
Videos