missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-2 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure rat IL-2 levels in cell culture supernates, serum, and plasma.

Brand:  R&D Systems™ SR2000

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462856

  • £1889.95 / Pack of 6


Estimated Shipment
28-02-2019
Add to basket

Description

Description

It contains E. coli-expressed recombinant rat IL-2 and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate the recombinant factor accurately. Results obtained using natural rat IL-2 showed dose response curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural rat IL-2
Specifications

Specifications

HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates, Serum, EDTA Plasma, Heparin Plasma
Rat
Sufficient materials to run ELISAs on at least six 96-well plates
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
116562
6 Plates
15pg/mL
Cell Culture Supernates (50μL), Serum (25μL), EDTA Plasma (25μL), Heparin Plasma (25μL)
Natural and recombinant rat IL-2
Videos