missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Rat IL-17A Platinum ELISA Kit

For the quantitative detection of rat IL-17A.

Brand:  Invitrogen™ BMS635

Code : Z2

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13485436

  • £352.87 / Each


EU Stock 13
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat IL-17A
Biotin-Conjugate anti-rat IL-17A monoclonal antibody
Streptavidin-HRP
Rat IL-17A Standard lyophilized, 200pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Rat
Interleukin 17 A, CTLA8
ELISA
1.0pg/mL
50μL
IL-17A
Videos

For Research Use Only