Thermo Scientific™ Pierce™ Rat IL-10 ELISA Kits

Measure levels of rat interleukin-10 in rat serum, plasma (EDTA, heparin, sodium citrate) and cell culture supernatant samples using 96-well plates.

Overview
Brand: Thermo Scientific™ Pierce™

Manufacturer Part Number: ERIL102

RAT IL-10 ELISA

UNSPSC: 41116126

Code: PN

Additional Details:
Additional Details: Weight: 0.10000kg



Product Code. 10200074

Quantity Price
1 £ 453.0 / 1 set
Estimated Shipment date
from Supplier 30-01-2017
Add to basket


Description and Specification

Specification

Sensitivity <3pg/mL
Formulation Complete ELISA kit with coated 96-well plates
Measurement Absorbance at 450nm minus absorbance at 550nm
Kit Contents 30X Wash Buffer, 2 x 50mL
Assay Range 16 to 500pg/mL
Sufficient For 2 x 96 microplate wells
Format Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Specificity No significant cross-reaction with rat IFNg, IL-1a, IL-1ß, IL-2, IL-4, IL-6, MCP-1, MIP-1a, RANTES, TNFa, human IL-10, or mouse IL-10
Description Rat IL-10 ELISA Kit
Sample Size 50µL
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Target Natural and recombinant rat interleukin-10 (IL-10)
Precision Dependent upon sample type (generally less than 10% CV)
Total Assay Time 3 hr.
Quantity 2 plate kit

These rat IL-10 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-10 (IL-10) in samples at 50μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution.The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interleukin-10 (IL-10)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 16 to 500pg/mL

  • Sensitivity: < 3pg/mL

  • Precision: Dependent upon sample type (generally less than 10% CV)

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IFNγ, IL-1α, IL-1β, IL-2, IL-4, IL-6, MCP-1, MIP-1α, RANTES, TNFα, human IL-10, or mouse IL-10

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Documentation

Technical documents