missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit, R&D Systems™

Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SRLB00

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442866

  • £2222.64 / Pack of 6

Estimated Shipment date
from Supplier 16-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Rat IL-1 beta/IL-1F2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat IL-1 beta/IL-1F2.
Specifications

Specifications

HRP
ELISA
Solid Phase Sandwich ELISA
1/Pk.
Cell Culture Supernates, Serum, EDTA Plasma
Rat
Store the unopened product at 2 - 8° C. Do not use past expiration date.
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
24494
6 Plates
5pg/mL
Cell Culture Supernates (50μL), Serum (17μL), EDTA Plasma (17μL)
Natural and recombinant rat IL-1 beta
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents