Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Isotype Control
£129.00 - £292.00
Specifications
Antigen | Rat IgG1 kappa |
---|---|
Clone | eBRG1 |
Conjugate | Super Bright 645 |
Form | Liquid |
Host Species | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15855683
|
Affymetrix eBioscience
64-4301-82 |
100 μg |
£292.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15845683
|
Invitrogen
64-4301-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in immunohistochemistry, immunocytochemistry, flow cytometric analysis, and ELISA. Applications Tested: Rat IgG1 Isotype Control has been tested by flow cytometric analysis of mouse splenocyte suspensions. It should be used at the same concentration as the experimental antibody. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Rat IgG1 kappa Control antibody specifically detects Rat IgG1 kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG1 kappa | |
Super Bright 645 | |
Rat | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eBRG1 | |
Liquid | |
RUO | |
IgG1 κ | |
Affinity chromatography |