missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgG (Total) Rat Uncoated ELISA Kit

For quantitative detection of rat total IgG.

Brand:  Invitrogen™ 88-50490-88

245.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15500927

  • £368.00 / Pack of 10
In Stock
to see stock.
Add to basket

Includes: Capture Antibody Concentrate

Description

Description

The Rat Total Immunoglobulin G (IgG) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat total IgG. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications

Specifications

1.6-100 ng/mL
HRP
Plasma, Serum, Supernatant
ELISA
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
10 x 96 Tests
Plasma, 10 μL; Serum, 10 μL; Supernatant, 50 μL
Immunoglobulin G
1 hr. 45 min.
1.6 ng/mL
ELISA Kit
Rat
Colorimetric Microplate Reader
HRP
RUO
2°C to 8°C
Rat
24 hr. 30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only