Learn More
RASA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557247
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PDDYSNFVIE DSVTTFKTIQ QIKSIIEKLD EPHEKYRKKR SSSAKYGSKE NPIVGKAS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
The Ras-related low molecular mass GTPases participate in signal transduction involving a variety of cellular functions, including cell-cycle progression, cellular differentiation, cytoskeletal organization, protein transport and secretion. The Ras-GTPase-activating protein (RASGAP) forms a complex with a second protein, p190, in growth factor stimulated and tyrosinekinase transformed cells. The RASGAP p190 complex may serve to couple Ras- and Rho-mediated signaling pathways. RASGAP forms an abundant SH2-mediated complex with p190 RhoGAP in cells expressing activated tyrosine kinases.Specifications
RASA2 | |
Polyclonal | |
Unconjugated | |
RASA2 | |
5430433H21Rik; AA517451; AU023900; CMAVM; CM-AVM; DKFZp434N071; GAP; GAP1M; GTPase-activating protein 1m; GTPase-activating protein of RAS; p120GAP; p120RASGAP; PKWS; ras GTPase-activating protein 2; RAS p21 protein activator 2; RASA; Rasa2; RASGAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5922 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.14 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15283 | |
RASA2 | |
Recombinant protein corresponding to Human RASA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only